Learn More
Medchemexpress LLC HY-19778A 1mg Medchemexpress, BPO-27 (racemate) CAS:1314873-02-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19778A1MG
Encompass_Preferred
Medchemexpress, HY-19778A 1mg BPO-27 (racemate) CAS:1314873-02-3 BPO-27 racemate is a potent CFTR inhibitor with an IC50 of 8 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0885