Learn More
IDEX Health & Science Upchurch BPR ASMB SS HLD GLD CAT 500PSI
Catalog No. NC1920904
Encompass_Preferred
Idex Back Pressure Regulators and Accessories, Stainless Steel, Pressure: 500 psi, Swept Volume: 129 uL, Back Pressure Regulator Assembly
Catalog No. NC1920904 Supplier IDEX Health & Science Upchurch Supplier No. U609
Idex Back Pressure Regulators and Accessories, Stainless Steel, Pressure: 500 psi, Swept Volume: 129 uL, Back Pressure Regulator Assembly