Learn More
IDEX Health & Science Upchurch BPR ASSEMBLY 20PSI W/CARTRIDGE
Catalog No. NC1920906
Encompass_Preferred
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 20 psi, Swept Volume: 134 uL, Back Pressure Regulator Assembly
Catalog No. NC1920906 Supplier IDEX Health & Science Upchurch Supplier No. P791
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 20 psi, Swept Volume: 134 uL, Back Pressure Regulator Assembly