Learn More
IDEX Health & Science Upchurch BPR ASSEMBLY 5PSI W/CARTRIDGE
Catalog No. NC1920905
Encompass_Preferred
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 5 psi, Swept Volume: 134 uL, Back Pressure Regulator Assembly
Catalog No. NC1920905 Supplier IDEX Health & Science Upchurch Supplier No. P790
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 5 psi, Swept Volume: 134 uL, Back Pressure Regulator Assembly