Learn More
IDEX Health & Science Upchurch BPR ASSMBLY BIOCOMPAT 100 PSI
Catalog No. NC1920896
Encompass_Preferred
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 100 psi, Swept Volume: 131 uL, Back Pressure Regulator Assembly
Catalog No. NC1920896 Supplier IDEX Health & Science Upchurch Supplier No. P787
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 100 psi, Swept Volume: 131 uL, Back Pressure Regulator Assembly