Learn More
IDEX Health & Science Upchurch BPR ASSMBLY BIOCOMPAT 250 PSI
Catalog No. NC1920898
Encompass_Preferred
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 250 psi, Swept Volume: 109 uL, Back Pressure Regulator Assembly
Catalog No. NC1920898 Supplier IDEX Health & Science Upchurch Supplier No. P788
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 250 psi, Swept Volume: 109 uL, Back Pressure Regulator Assembly