Learn More
IDEX Health & Science Upchurch BPR ASSMBLY BIOCOMPAT 40 PSI
Catalog No. NC1920893
Encompass_Preferred
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 40 psi, Swept Volume: 131 uL, Back Pressure Regulator Assembly
Catalog No. NC1920893 Supplier IDEX Health & Science Upchurch Supplier No. P785
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 40 psi, Swept Volume: 131 uL, Back Pressure Regulator Assembly