Learn More
IDEX Health & Science Upchurch BPR ASSMBLY BIOCOMPAT 500 PSI
Catalog No. NC1920899
Encompass_Preferred
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 500 psi, Swept Volume: 96 uL, Back Pressure Regulator Assembly
Catalog No. NC1920899 Supplier IDEX Health & Science Upchurch Supplier No. P789
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 500 psi, Swept Volume: 96 uL, Back Pressure Regulator Assembly