Learn More
IDEX Health & Science Upchurch BPR ASSMBLY BIOCOMPAT 75 PSI
Catalog No. NC1920894
Encompass_Preferred
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 75 psi, Swept Volume: 131 uL, Back Pressure Regulator Assembly
Catalog No. NC1920894 Supplier IDEX Health & Science Upchurch Supplier No. P786
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 75 psi, Swept Volume: 131 uL, Back Pressure Regulator Assembly