Learn More
IDEX Health & Science Upchurch BPR CART GOLD COATED 500 PSI
Catalog No. NC1920892
Encompass_Preferred
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 500 psi, Swept Volume: 89 uL, Back Pressure Regulator Assembly
Catalog No. NC1920892 Supplier IDEX Health & Science Upchurch Supplier No. P765
Idex Back Pressure Regulators and Accessories, PEEK, Pressure: 500 psi, Swept Volume: 89 uL, Back Pressure Regulator Assembly