Learn More
IDEX Health & Science Upchurch BPR CART GOLD COATED 75 PSI
Catalog No. NC1920889
Encompass_Preferred
Idex Back Pressure Regulators: Cartridges, Pressure: 75 psi, Swept Volume: 125 uL, For Use With: Back Pressure Regulators
Catalog No. NC1920889 Supplier IDEX Health & Science Upchurch Supplier No. P762
Idex Back Pressure Regulators: Cartridges, Pressure: 75 psi, Swept Volume: 125 uL, For Use With: Back Pressure Regulators