Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BPR1J-097 hydrochloride | 00-00-0 | 99.2% | C27H29ClN6O3S | 10MG
SDP

Catalog No. 5000259703
Encompass_Preferred

BPR1J-097 hydrochloride is a small-molecule FLT3 kinase inhibitor with nanomolar potency for use in biochemical and cellular research. Supplied as the hydrochloride salt with high chemical purity, it is intended for pharmacological profiling and in vitro assay applications where selective FLT3 inhibition is required.

  • Potent FLT3 inhibition (IC50 ~11 nM).
  • High chemical purity (99.2%).
  • Hydrochloride salt form for improved solubility.
  • Characterized by molecular formula C27H29ClN6O3S and MW 553.08.
  • Intended for research use in biochemical and cellular assays.

Catalog No. 50-002-59703 Supplier Medchemexpress LLC Supplier No. HY13537A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BPR1J-097 hydrochloride is a small-molecule FLT3 kinase inhibitor with nanomolar potency for use in biochemical and cellular research. Supplied as the hydrochloride salt with high chemical purity, it is intended for pharmacological profiling and in vitro assay applications where selective FLT3 inhibition is required.

  • Potent FLT3 inhibition (IC50 ~11 nM).
  • High chemical purity (99.2%).
  • Hydrochloride salt form for improved solubility.
  • Characterized by molecular formula C27H29ClN6O3S and MW 553.08.
  • Intended for research use in biochemical and cellular assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.