Learn More
Medchemexpress LLC BPR1M97 | 2059904-66-2 | 99.4% | 10 MG
SDP
Catalog No. 5000338636
Encompass_Preferred
BPR1M97 is a dual-acting mu opioid receptor (MOP) and nociceptin-orphanin FQ peptide (NOP) receptor agonist. It exhibits high potency, penetrates the blood-brain barrier, and produces potent antinociceptive effects.
  • Dual-acting mu opioid receptor (MOP) agonist
  • Nociceptin-orphanin FQ peptide (NOP) receptor agonist
  • High potency
  • Penetrates blood-brain barrier
  • Produces potent antinociceptive effects
  • For research use only
Catalog No. 50-003-38636 Supplier Medchemexpress LLC Supplier No. HY12886510MG
BPR1M97 is a dual-acting mu opioid receptor (MOP) and nociceptin-orphanin FQ peptide (NOP) receptor agonist. It exhibits high potency, penetrates the blood-brain barrier, and produces potent antinociceptive effects.
  • Dual-acting mu opioid receptor (MOP) agonist
  • Nociceptin-orphanin FQ peptide (NOP) receptor agonist
  • High potency
  • Penetrates blood-brain barrier
  • Produces potent antinociceptive effects
  • For research use only