Learn More
Medchemexpress LLC BPR1M97 | 2059904-66-2 | 99.4% | 100 MG
SDP
Catalog No. 5000338965
Encompass_Preferred
BPR1M97 is a dual-acting mu opioid receptor (MOP) and nociceptin-orphanin FQ peptide (NOP) receptor agonist. It exhibits high potency, blood-brain barrier penetration, and produces potent antinociceptive effects. This compound is intended for research use only.
  • Dual-acting mu opioid receptor (MOP) agonist
  • Dual-acting nociceptin-orphanin FQ peptide (NOP) receptor agonist
  • High potency
  • Penetrates the blood-brain barrier
  • Produces potent antinociceptive effects
  • Ki values: 1.8 nM for MOP, 4.2 nM for NOP
  • Agonist activity at human MOR: IC50 0.05 μM
  • Agonist activity at human KOR: IC50 0.44 μM
Catalog No. 50-003-38965 Supplier Medchemexpress LLC Supplier No. HY128865100MG
BPR1M97 is a dual-acting mu opioid receptor (MOP) and nociceptin-orphanin FQ peptide (NOP) receptor agonist. It exhibits high potency, blood-brain barrier penetration, and produces potent antinociceptive effects. This compound is intended for research use only.
  • Dual-acting mu opioid receptor (MOP) agonist
  • Dual-acting nociceptin-orphanin FQ peptide (NOP) receptor agonist
  • High potency
  • Penetrates the blood-brain barrier
  • Produces potent antinociceptive effects
  • Ki values: 1.8 nM for MOP, 4.2 nM for NOP
  • Agonist activity at human MOR: IC50 0.05 μM
  • Agonist activity at human KOR: IC50 0.44 μM