Learn More
Medchemexpress LLC BPR1M97 | 2059904-66-2 | 99.4% | 50 MG
SDP
Catalog No. 5000340062
Encompass_Preferred
BPR1M97 is a dual-acting mu opioid receptor (MOP) and nociceptin-orphanin FQ peptide (NOP) receptor agonist. It has Ki values of 1.8 nM for MOP and 4.2 nM for NOP. BPR1M97 exhibits high potency, penetrates the blood-brain barrier, and produces potent antinociceptive effects. It is intended for research use only and is not sold to patients.
  • Dual-acting mu opioid receptor (MOP) and nociceptin-orphanin FQ peptide (NOP) receptor agonist
  • Ki values of 1.8 nM for MOP and 4.2 nM for NOP
  • High potency
  • Effective blood-brain barrier penetration
  • Potent antinociceptive effects
  • Intended for research use only
Catalog No. 50-003-40062 Supplier Medchemexpress LLC Supplier No. HY12886550MG
BPR1M97 is a dual-acting mu opioid receptor (MOP) and nociceptin-orphanin FQ peptide (NOP) receptor agonist. It has Ki values of 1.8 nM for MOP and 4.2 nM for NOP. BPR1M97 exhibits high potency, penetrates the blood-brain barrier, and produces potent antinociceptive effects. It is intended for research use only and is not sold to patients.
  • Dual-acting mu opioid receptor (MOP) and nociceptin-orphanin FQ peptide (NOP) receptor agonist
  • Ki values of 1.8 nM for MOP and 4.2 nM for NOP
  • High potency
  • Effective blood-brain barrier penetration
  • Potent antinociceptive effects
  • Intended for research use only