Learn More
TARGETMOL CHEMICALS INC BPR1M97 5MG
SDP
Catalog No. 502581704
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BPR1M97 is a mu opioid receptor (MOP) and neuropeptide-orphin FQ (NOP) receptor agonist with blood-brain barrier permeability and potency with Kis values of 1.8 nM for MOP and 4.2 nM for NOP.BPR1M97 exhibits anti-injurious effects and antinociceptive effects. purity: 99%
Catalog No. 50-258-1704 Supplier TARGETMOL CHEMICALS INC Supplier No. T105935MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BPR1M97 is a mu opioid receptor (MOP) and neuropeptide-orphin FQ (NOP) receptor agonist with blood-brain barrier permeability and potency with Kis values of 1.8 nM for MOP and 4.2 nM for NOP.BPR1M97 exhibits anti-injurious effects and antinociceptive effects. purity: 99%