Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ BPRBAND Bump Cap Sweat Band

Catalog No. 19155228 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-155-228 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-155-228 Supplier Pyramex™ Supplier No. BPRBAND
This item is not returnable. View return policy

Sweat band for bump caps

Replacement sweat band

Specifications

Color Black
For Use With (Equipment) Bump Cap
Product Type Sweatband
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.