Learn More
Higgins Analytical Inc BPSPN PROTO MACRO 300 C18/20BX
Catalog No. NC1479795
Encompass
bioPureSPN Macro, PROTO 300 C18 (Green) 35-350µg capacity, 0.4ml, 100mg, 300Å, 20/bx
Catalog No. NC1479795 Supplier Higgins Analytical Inc Supplier No. HMM S18V.20
bioPureSPN Macro, PROTO 300 C18 (Green) 35-350µg capacity, 0.4ml, 100mg, 300Å, 20/bx