Learn More
Medchemexpress LLC HY-12683 10mg Medchemexpress, BPTES CAS:314045-39-1 Purity:>98%
SDP
Catalog No. 502028909
Encompass_Preferred
Medchemexpress, HY-12683 10mg BPTES CAS:314045-39-1 BPTES is an allosteric and selective glutaminase inhibitor with an IC50 of 0.16 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8909 Supplier Medchemexpress LLC Supplier No. HY1268310MG
Medchemexpress, HY-12683 10mg BPTES CAS:314045-39-1 BPTES is an allosteric and selective glutaminase inhibitor with an IC50 of 0.16 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.