Learn More
Selleck Chemical LLC BPTES 10mg 314045-39-1
SDP
Supplier:  Selleck Chemical LLC S775310MG
Encompass_Preferred
BPTES is a potent and selective Glutaminase GLS1 (KGA) inhibitor with IC50 of 0.16 ?M. It has no effect on glutamate dehydrogenase activity and causes only a very slight inhibition of ?-glutamyl transpeptidase activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1099646