Learn More
Medchemexpress LLC BPTES | 314045-39-1 | 98.0% | 200 MG
SDP
Catalog No. 5000342238
Encompass_Preferred
BPTES is an allosteric and selective glutaminase inhibitor with an IC50 of 0.16 μM. It is for research use only.
  • Inhibits glutaminase activity
  • Diminishes glutamate and α-KG levels
  • Increases glycolytic intermediates
  • Inhibits PDAC cell proliferation
  • Slows growth of mutant IDH1 cells
  • Shows synergistic anti-cancer effect with 5-FU in A549 and EKVX cell lines
  • Reduces growth in most NSCLC cell lines
  • Reduces levels of TCA cycle metabolites
  • Reduces ATP production
  • Attenuates tumor growth in patient-derived pancreatic orthotopic tumor models
Catalog No. 50-003-42238 Supplier Medchemexpress LLC Supplier No. HY12683200MG
BPTES is an allosteric and selective glutaminase inhibitor with an IC50 of 0.16 μM. It is for research use only.
  • Inhibits glutaminase activity
  • Diminishes glutamate and α-KG levels
  • Increases glycolytic intermediates
  • Inhibits PDAC cell proliferation
  • Slows growth of mutant IDH1 cells
  • Shows synergistic anti-cancer effect with 5-FU in A549 and EKVX cell lines
  • Reduces growth in most NSCLC cell lines
  • Reduces levels of TCA cycle metabolites
  • Reduces ATP production
  • Attenuates tumor growth in patient-derived pancreatic orthotopic tumor models