Learn More
Medchemexpress LLC HY-132889 5mg , BPTF-IN-BZ1 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1328895MG
Encompass_Preferred
HY-132889 5mg BPTF-IN-BZ1 CAS: BPTF-IN-BZ1, a BPTF inhibitor, possesses a high potency (Kd = 6.3 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1758