Learn More
TARGETMOL CHEMICALS INC BPTU 10MG
SDP
Catalog No. 502576801
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BPTU (BMS-646786) is an allosteric antagonist of P2Y1 (EC50 = 0.06-0.3 uM). Non-nucleotide ligand. Binds receptor outside of the helical bundle. Blocks inhibition of spontaneous contraction of rat and mouse colon induced by electrical field stimulation nicotine and P2Y agonists. Antithrombotic; reduces platelet aggregation. purity: 99%
Catalog No. 50-257-6801 Supplier TARGETMOL CHEMICALS INC Supplier No. T413210MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BPTU (BMS-646786) is an allosteric antagonist of P2Y1 (EC50 = 0.06-0.3 uM). Non-nucleotide ligand. Binds receptor outside of the helical bundle. Blocks inhibition of spontaneous contraction of rat and mouse colon induced by electrical field stimulation nicotine and P2Y agonists. Antithrombotic; reduces platelet aggregation. purity: 99%