Learn More
New England Biolabs, Inc. Bpu10I – 200 units
SDP
Catalog No. 50994929
Encompass
A restriction endonuclease that recognizes the sequence CC^TNA_GC.
Catalog No. 50-994-929 Supplier New England Biolabs, Inc. Supplier No. R0649S
A restriction endonuclease that recognizes the sequence CC^TNA_GC.