Learn More
Medchemexpress LLC BPU17 | 2198977-68-1 | 98.2% | 252.70 | 250 MG
SDP
Catalog No. 5000392558
Encompass_Preferred
BPU17 binds to PHB1, causing mild defects in mitochondrial function through impaired PHB1-PHB2 interaction. This inhibits SRF/CArG-box-dependent transcription, suppressing the epithelial-mesenchymal transition (EMT) of retinal pigment epithelial cells (RPEs). BPU17 also shows antifibrotic activity in vivo and is promising for research into anti-neovascular age-related macular degeneration (nAMD) agents.
  • EMT inhibitor
  • PHB1 activator
  • Antifibrotic activity in vivo
  • Promising for research of anti-neovascular age-related macular degeneration (nAMD) agent
  • Solid, white to off-white appearance
Catalog No. 50-003-92558 Supplier Medchemexpress LLC Supplier No. 250MGHY162904
BPU17 binds to PHB1, causing mild defects in mitochondrial function through impaired PHB1-PHB2 interaction. This inhibits SRF/CArG-box-dependent transcription, suppressing the epithelial-mesenchymal transition (EMT) of retinal pigment epithelial cells (RPEs). BPU17 also shows antifibrotic activity in vivo and is promising for research into anti-neovascular age-related macular degeneration (nAMD) agents.
  • EMT inhibitor
  • PHB1 activator
  • Antifibrotic activity in vivo
  • Promising for research of anti-neovascular age-related macular degeneration (nAMD) agent
  • Solid, white to off-white appearance