Learn More
New England Biolabs, Inc. BpuEI – 500 units
SDP
Catalog No. 50812380
Encompass
A restriction endonuclease that recognizes the sequence CTTGAGNNNNNNNNNNNNNN_NN^.
Catalog No. 50-812-380 Supplier New England Biolabs, Inc. Supplier No. R0633S
A restriction endonuclease that recognizes the sequence CTTGAGNNNNNNNNNNNNNN_NN^.