Learn More
AdipoGen bpV(bipy) (5 mg)
SDP
Supplier:  AdipoGen AGCR10047M005
Encompass_Preferred
Chemical. CAS: 1173697-60-3 | 42494-71-3 (anhydrous). Formula: K[VO(O2)2C10H8N2] . 3H2O. MW: 326.2 . 54.0. Potent protein phosphotyrosine phosphatase inhibitor. Insulin mimetic in vitro and in vivo. PTEN inhibitor.
Catalog No. 50-205-3803