Learn More
Enzo Life Sciences bpV(HOpic) (25mg)
SDP
Catalog No. 502008426
Encompass_Preferred
PTP inhibitor, PTEN Inhibitor. Protein phosphotyrosine phosphatase inhibitor. Formula: K2[VO(O2)2C6H4NO3]. MW: 347.2. Purity: ≥95% (HPLC). Appearance: Yellow crystalline solid. Solubility: Soluble in DMSO or water. Note: This product is not stable in water. Starts decomposition immediately after solubilization. Rate of decomposition is slower in dry DMSO. Long Term Storage: -20°C. Handling: Protect from light and moisture.
Catalog No. 50-200-8426 Supplier Enzo Life Sciences Supplier No. ALX270206M025
PTP inhibitor, PTEN Inhibitor. Protein phosphotyrosine phosphatase inhibitor. Formula: K2[VO(O2)2C6H4NO3]. MW: 347.2. Purity: ≥95% (HPLC). Appearance: Yellow crystalline solid. Solubility: Soluble in DMSO or water. Note: This product is not stable in water. Starts decomposition immediately after solubilization. Rate of decomposition is slower in dry DMSO. Long Term Storage: -20°C. Handling: Protect from light and moisture.