Learn More
Selleck Chemical LLC bpV (HOpic) 5mg 722494-26-0 Bisperoxovanadium (HOpic)
SDP
Catalog No. 501948142
Encompass_Preferred
bpV (HOpic) (Bisperoxovanadium (HOpic)) is a potent inhibitor of PTEN with an IC50 of 14 nM. The IC50s for PTP-? and PTP-1B are about 350- and 1800-fold higher than the IC50 for PTEN, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8142 Supplier Selleck Chemical LLC Supplier No. S86515MG
bpV (HOpic) (Bisperoxovanadium (HOpic)) is a potent inhibitor of PTEN with an IC50 of 14 nM. The IC50s for PTP-? and PTP-1B are about 350- and 1800-fold higher than the IC50 for PTEN, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.