Learn More
Enzo Life Sciences bpV(HOpic) (5mg)
SDP
Catalog No. 502008425
Encompass_Preferred
Protein phosphotyrosine phosphatase inhibitor. Formula: K2[VO(O2)2C6H4NO3]. MW: 347.2. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO or water. Note: This product is not stable in water. Starts decomposition immediately after solubilization. Rate of decomposition is slower in dry DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8425 Supplier Enzo Life Sciences Supplier No. ALX270206M005
Protein phosphotyrosine phosphatase inhibitor. Formula: K2[VO(O2)2C6H4NO3]. MW: 347.2. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO or water. Note: This product is not stable in water. Starts decomposition immediately after solubilization. Rate of decomposition is slower in dry DMSO. Long Term Storage: -20°C.