Learn More
AdipoGen bpV(pic) (5 mg)
SDP
Supplier:  AdipoGen AGCR10043M005
Encompass_Preferred
Chemical. CAS: 148556-27-8. Formula: K2 [VO(O2)2C6H4NO2] . 2 H2O. MW: 331.2 . 36.0. Potent protein phosphotyrosine phosphatase inhibitor. Insulin mimetic. Angiogenesis inhibitor. PTEN inhibitor.
Catalog No. 50-205-3800