Learn More
AdipoGen bpVHOpic
SDP
Catalog No. 502562378
Encompass_Preferred
Chemical. CAS 722494-26-0. Formula K2VOO22C6H4NO3. MW 347.2. Potent protein phosphotyrosine phosphatase inhibitor. Insulin mimetic. PTEN inhibitor.
Catalog No. 50-256-2378 Supplier AdipoGen Supplier No. AGCR10044M025
Chemical. CAS 722494-26-0. Formula K2VOO22C6H4NO3. MW 347.2. Potent protein phosphotyrosine phosphatase inhibitor. Insulin mimetic. PTEN inhibitor.