Learn More
TARGETMOL CHEMICALS INC BPVHOPIC 5MG
SDP
Catalog No. 502579105
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BpV(HOpic) (bpV (HOpic)) is a potent inhibitor of PTEN (IC50 of 14 nM). purity: 99%
Catalog No. 50-257-9105 Supplier TARGETMOL CHEMICALS INC Supplier No. T79535MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BpV(HOpic) (bpV (HOpic)) is a potent inhibitor of PTEN (IC50 of 14 nM). purity: 99%