Learn More
AdipoGen bpVphen
SDP
Catalog No. 502562381
Encompass_Preferred
Chemical. CAS 171202-16-7. Formula KVOO22C12H8N2 . 3H2O. MW 350.2 . 54.0. Potent protein phosphotyrosine phosphatase inhibitor. Insulin receptor kinase IRK inducer. Insulin mimetic in vitro and in vivo. Apoptosis inducer. ERK inducer. Mitogen-activated protein kinase phosphatase-1 MKP-1 inducer. Angiogenesis inhibitor. PTEN inhibitor.
Catalog No. 50-256-2381 Supplier AdipoGen Supplier No. AGCR10042M005
Chemical. CAS 171202-16-7. Formula KVOO22C12H8N2 . 3H2O. MW 350.2 . 54.0. Potent protein phosphotyrosine phosphatase inhibitor. Insulin receptor kinase IRK inducer. Insulin mimetic in vitro and in vivo. Apoptosis inducer. ERK inducer. Mitogen-activated protein kinase phosphatase-1 MKP-1 inducer. Angiogenesis inhibitor. PTEN inhibitor.