Learn More
AdipoGen BQ-123 sodium salt
SDP
Catalog No. 502562465
Encompass_Preferred
Chemical. CAS 136655-57-7. Formula C31H42N6O7 . Na. MW 609.7 . 23.0. Synthetic. Potent and selective endothelin receptor A ETA antagonist. Suppresses the ET-1-induced decrease of LPL activity. Shows protective effects in ischemic acute renal failure. Neuroprotective. Anti-hypertensive. Inhibits ET-1 receptor binding and blocks Ca2 mobilization, cellular contraction and MAP kinase activation. Cardiovascular agent. Reduces myocardial infarct size and oxidant injury. Selected Reviews.
Catalog No. 50-256-2465 Supplier AdipoGen Supplier No. AGCP30001M001
Chemical. CAS 136655-57-7. Formula C31H42N6O7 . Na. MW 609.7 . 23.0. Synthetic. Potent and selective endothelin receptor A ETA antagonist. Suppresses the ET-1-induced decrease of LPL activity. Shows protective effects in ischemic acute renal failure. Neuroprotective. Anti-hypertensive. Inhibits ET-1 receptor binding and blocks Ca2 mobilization, cellular contraction and MAP kinase activation. Cardiovascular agent. Reduces myocardial infarct size and oxidant injury. Selected Reviews.