Learn More
Enzo Life Sciences ET B receptor antagonist. sodium salt (1mg). CAS: 156161-89-6
SDP
Catalog No. 502007808
Encompass_Preferred
Highly potent and selective ETB-receptor antagonist. Alternative name: BQ-788 . Na. Formula: C34H50N5O7Na. MW: 663.7. Purity: ≥95% (HPLC). Formulation: Lyophilized from 0.1M sodium bicarbonate. Solubility: Soluble in methanol (0.5mg/ml), DMSO (4mg/ml), 0.1M sodium bicarbonate (3mg/ml) or 100% ethanol (4mg/ml); insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7808 Supplier Enzo Life Sciences Supplier No. ALX155020M001
Highly potent and selective ETB-receptor antagonist. Alternative name: BQ-788 . Na. Formula: C34H50N5O7Na. MW: 663.7. Purity: ≥95% (HPLC). Formulation: Lyophilized from 0.1M sodium bicarbonate. Solubility: Soluble in methanol (0.5mg/ml), DMSO (4mg/ml), 0.1M sodium bicarbonate (3mg/ml) or 100% ethanol (4mg/ml); insoluble in water. Long Term Storage: -20°C.