Learn More
AdipoGen BQ-788 . sodium salt,Chemical. CAS: 156161-89-6
SDP
Supplier:  AdipoGen AGCP30012M001
Encompass_Preferred
BQ-788 . sodium salt, AdipoGen Life Sciences. Chemical. CAS: 156161-89-6. Formula: C34H50N5O7 . Na. MW: 640.8 . 23.0. Synthetic. Potent and selective endothelin B receptor (ETBR) antagonist.
Catalog No. 50-101-6216