Learn More
AdipoGen BQ-788 sodium salt
SDP
Catalog No. 502562451
Encompass_Preferred
Chemical. CAS 156161-89-6. Formula C34H50N5O7 . Na. MW 640.8 . 23.0. Synthetic. Potent and selective endothelin B receptor ETBR antagonist.
Catalog No. 50-256-2451 Supplier AdipoGen Supplier No. AGCP30012C500
Chemical. CAS 156161-89-6. Formula C34H50N5O7 . Na. MW 640.8 . 23.0. Synthetic. Potent and selective endothelin B receptor ETBR antagonist.