Learn More
Medchemexpress LLC HY-101858 5mg Medchemexpress, BQCA CAS:338747-41-4 Purity:>98%
SDP
Catalog No. 501967893
Encompass_Preferred
Medchemexpress, HY-101858 5mg BQCA CAS:338747-41-4 BQCA a highly selective allosteric modulator of the M1 mAChR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7893 Supplier Medchemexpress LLC Supplier No. HY1018585MG
Medchemexpress, HY-101858 5mg BQCA CAS:338747-41-4 BQCA a highly selective allosteric modulator of the M1 mAChR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.