Learn More
Apexbio Technology LLC BQU57, 5mg. Cas: 1637739-82-2 MFCD: NA. Derivative of RBC8
SDP
Supplier:  Apexbio Technology LLC A95035
Encompass_Preferred
BQU57 is a derivative of RBC8. It show selectivity for Ral relative to the GTPases Ras and RhoA and inhibit tumour xenograft growth. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5197