Learn More
C&G Containers BR 8OZ 24-400 BLKPHENPS 108/CS
SDP
Catalog No. NC9498832
Encompass_Preferred
8oz Flint Boston Round 24-400 Black Phenolic PS, assembled only, bottles with cap attached L3 108/cs
Catalog No. NC9498832 Supplier C&G Containers Supplier No. LFB0089005
8oz Flint Boston Round 24-400 Black Phenolic PS, assembled only, bottles with cap attached L3 108/cs