Learn More
Medchemexpress LLC Br-Boc-C2-azido | 1120364-53-5 | >98% | 236.07 | 1 G
SDP
Catalog No. 5000349303
Encompass_Preferred
Br-Boc-C2-azido is an alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent due to its Azide group, enabling participation in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Alkyl/ether-based linker for PROTAC synthesis
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Supports PROTACs in degrading target proteins via the ubiquitin-proteasome system
Catalog No. 50-003-49303 Supplier Medchemexpress LLC Supplier No. HY1384641G
Br-Boc-C2-azido is an alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent due to its Azide group, enabling participation in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Alkyl/ether-based linker for PROTAC synthesis
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Supports PROTACs in degrading target proteins via the ubiquitin-proteasome system