Learn More
Medchemexpress LLC Br-PEG3-ethyl acetat 100mg
SDP
Catalog No. 5000205334
Encompass_Preferred
Br-PEG3-ethyl acetate is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05334 Supplier Medchemexpress LLC Supplier No. HYW096139100MG
Br-PEG3-ethyl acetate is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]