Learn More
Medchemexpress LLC Br-PEG3-ethyl acetat 25mg
SDP
Catalog No. 5000204559
Encompass_Preferred
Br-PEG3-ethyl acetate is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-04559 Supplier Medchemexpress LLC Supplier No. 25MGHYW096139
Add to Cart
Add to Cart
Br-PEG3-ethyl acetate is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]