Learn More
Medchemexpress LLC Br-PEG3-ethyl acetat 50mg
SDP
Catalog No. 5000205722
Encompass_Preferred
Br-PEG3-ethyl acetate is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05722 Supplier Medchemexpress LLC Supplier No. HYW09613950MG
Br-PEG3-ethyl acetate is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]