Learn More
Medchemexpress LLC Br-PEG4-CH2-Boc | 1807505-29-8 | ≥97.0% | 100 MG
SDP
Catalog No. 5000339803
Encompass_Preferred
Br-PEG4-CH2-Boc is a PEG- and Alkyl/ether-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs utilize two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a specific target protein, to facilitate the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG- and alkyl/ether-based linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
  • For research use only
Catalog No. 50-003-39803 Supplier Medchemexpress LLC Supplier No. HY130536100MG
Br-PEG4-CH2-Boc is a PEG- and Alkyl/ether-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs utilize two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a specific target protein, to facilitate the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG- and alkyl/ether-based linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
  • For research use only