Learn More
Medchemexpress LLC Br-PEG4-CH2-Boc | 1807505-29-8 | ≥97.0% | 250 MG
SDP
Catalog No. 5000341965
Encompass_Preferred
Br-PEG4-CH2-Boc is a PEG- and Alkyl/ether-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs are molecules designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.
  • PEG- and alkyl/ether-based PROTAC linker
  • For use in PROTAC synthesis
  • PROTACs composed of two ligands connected by a linker
  • Targets E3 ubiquitin ligase and target proteins
  • Leverages the intracellular ubiquitin-proteasome system
  • Achieves selective degradation of target proteins
Catalog No. 50-003-41965 Supplier Medchemexpress LLC Supplier No. 250MGHY130536
Br-PEG4-CH2-Boc is a PEG- and Alkyl/ether-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs are molecules designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.
  • PEG- and alkyl/ether-based PROTAC linker
  • For use in PROTAC synthesis
  • PROTACs composed of two ligands connected by a linker
  • Targets E3 ubiquitin ligase and target proteins
  • Leverages the intracellular ubiquitin-proteasome system
  • Achieves selective degradation of target proteins