Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Br-PEG4-CH2-Boc | 1807505-29-8 | ≥97.0% | 5 G
SDP

Catalog No. 5000340556
Encompass_Preferred

Br-PEG4-CH2-Boc is a PEG- and Alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. These PROTACs feature two distinct ligands connected by a linker, one for an E3 ubiquitin ligase and another for the target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.

  • PEG- and Alkyl/ether-based linker
  • Facilitates PROTAC synthesis
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight of 371.26
  • Chemical formula: C14H27BrO6
  • Appears as a colorless to light yellow liquid
  • Purity of ≥97.0%
  • Relevant for cancer and cancer targeted therapy research

Catalog No. 50-003-40556 Supplier Medchemexpress LLC Supplier No. HY1305365G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Br-PEG4-CH2-Boc is a PEG- and Alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. These PROTACs feature two distinct ligands connected by a linker, one for an E3 ubiquitin ligase and another for the target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.

  • PEG- and Alkyl/ether-based linker
  • Facilitates PROTAC synthesis
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight of 371.26
  • Chemical formula: C14H27BrO6
  • Appears as a colorless to light yellow liquid
  • Purity of ≥97.0%
  • Relevant for cancer and cancer targeted therapy research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.