Learn More
Medchemexpress LLC Br-PEG4-OH | 85141-94-2 | 98.2% | 5 G
SDP
Catalog No. 5000338638
Encompass_Preferred
Br-PEG4-OH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two different ligands linked together; one targets an E3 ubiquitin ligase and the other targets the protein of interest. Their function is to leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Utilized in the synthesis of protacs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-38638 Supplier Medchemexpress LLC Supplier No. HY1305125G
Br-PEG4-OH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two different ligands linked together; one targets an E3 ubiquitin ligase and the other targets the protein of interest. Their function is to leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Utilized in the synthesis of protacs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins