Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Br-PEG4-THP | 1803547-60-5 | 341.24 | 100 MG
SDP

Catalog No. 5000349299
Encompass_Preferred

Br-PEG4-THP is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only and not sold to patients.

  • CAS number: 1803547-60-5
  • Molecular weight: 341.24
  • Chemical formula: C13H25BrO5
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Shipping conditions: Room temperature in continental US
  • Storage conditions for pure form: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-49299 Supplier Medchemexpress LLC Supplier No. HY138372100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Br-PEG4-THP is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only and not sold to patients.

  • CAS number: 1803547-60-5
  • Molecular weight: 341.24
  • Chemical formula: C13H25BrO5
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Shipping conditions: Room temperature in continental US
  • Storage conditions for pure form: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.